2-Methylthioanisole ≥95%

Supplier: AOBChem USA

Warning

27383-500MG 27383-1G 27383-5G 27383-25G
77324-872EA 93.73 USD
77324-872 77342-476 77361-196 77372-958
2-Methylthioanisole ≥95%
2-Methylthioanisole

Precaución: for Research Use Only

Formula: C₈H₁₀S
MW: 138.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00465079
CAS Number: 14092-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR