Potassium (2,5-dibromophenyl)trifluoroborate ≥95%

Supplier: AOBChem USA

Warning

27330-1G 27330-5G 27330-25G
77341-172EA 73.13 USD
77341-172 77361-192 77372-954
Potassium (2,5-dibromophenyl)trifluoroborate ≥95%
Potassium (2,5-dibromophenyl)trifluoroborate

Precaución: for Research Use Only

Formula: C₆H₃BBr₂F₃K+
MW: 341.8 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32215274
CAS Number: 1448312-02-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR