5-(Dimethylamino)-2-methylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

27250-500MG 27250-1G 27250-5G
77339-792EA 643.75 USD
77339-792 77348-710 77372-950
5-(Dimethylamino)-2-methylbenzoic acid ≥95%
5-(Dimethylamino)-2-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MW: 179.216 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20651077
CAS Number: 65399-11-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR