tert-Butyl(2-fluoro-5-methoxyphenoxy)dimethylsilane ≥97%

Supplier: AOBChem USA

Warning

27201-500MG 27201-1G 27201-5G 27201-25G
77324-866EA 101.97 USD
77324-866 77342-984 77361-184 77372-946
tert-Butyl(2-fluoro-5-methoxyphenoxy)dimethylsilane ≥97%
tert-Butyl(2-fluoro-5-methoxyphenoxy)dimethylsilane

Precaución: for Research Use Only

Formula: C₁₃H₂₁FO₂Si
MW: 256.389 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34178492
CAS Number: 1350551-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR