2,6,-Dimethyl-4-(1-methylethoxy)-benzenemethanol ≥97%

Supplier: AOBChem USA

Warning

27198-500MG 27198-1G 27198-5G 27198-25G
77324-864EA 222.48 USD
77324-864 77348-706 77361-182 77384-474
2,6,-Dimethyl-4-(1-methylethoxy)-benzenemethanol ≥97%
2,6,-Dimethyl-4-(1-methylethoxy)-benzenemethanol

Precaución: for Research Use Only

Formula: C₁₂H₁₈O₂
MW: 194.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28015442
CAS Number: 1506593-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR