4-Fluoro-2-isopropoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

27170-500MG 27170-1G 27170-5G 27170-25G
77324-862EA 154.5 USD
77324-862 77345-548 77361-180 77384-472
4-Fluoro-2-isopropoxybenzoic acid ≥97%
4-Fluoro-2-isopropoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₃
MW: 198.191 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28015441
CAS Number: 1369833-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR