3-Bromo-6-fluoro-2-methoxybenzenemethanol ≥97%

Supplier: AOBChem USA

Warning

27165-500MG 27165-1G 27165-5G 27165-25G
77324-860EA 176.13 USD
77324-860 77346-516 77361-178 77384-470
3-Bromo-6-fluoro-2-methoxybenzenemethanol ≥97%
3-Bromo-6-fluoro-2-methoxybenzenemethanol

Precaución: for Research Use Only

Formula: C₈H₈BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD28015439
CAS Number: 648439-20-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR