3-Cyclopropyl-2-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

27129-500MG 27129-1G 27129-5G 27129-25G
77324-858EA 252.35 USD
77324-858 77348-704 77361-176 77384-468
3-Cyclopropyl-2-methylpyridine ≥97%
3-Cyclopropyl-2-methylpyridine

Precaución: for Research Use Only

Formula: C₉H₁₁N
Storage Temperature: Ambient
MDL Number: MFCD26743572
CAS Number: 1346533-28-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR