3-Fluoro-2-(trimethylsilyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

27121-500MG 27121-1G 27121-5G
77336-232EA 384.19 USD
77336-232 77348-700 77372-940
3-Fluoro-2-(trimethylsilyl)phenylboronic acid ≥97%
3-Fluoro-2-(trimethylsilyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₄BFO₂Si
MW: 212.101 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18383410
CAS Number: 1257792-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR