6-Bromo-2,3-dihydro-benzo[1,4]dioxine-5-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

27097-500MG 27097-1G 27097-5G
77336-056EA 375.95 USD
77336-056 77348-698 77372-938
6-Bromo-2,3-dihydro-benzo[1,4]dioxine-5-carboxylic acid ≥97%
6-Bromo-2,3-dihydro-benzo[1,4]dioxine-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₇BrO₄
MW: 259.053 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902570
CAS Number: 69464-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR