5-Bromo-4-methyl-N,N-dimethylpyridine-2-carboxamide ≥95%

Supplier: AOBChem USA

Warning

27058-500MG 27058-1G 27058-5G 27058-25G
77324-850EA 176.13 USD
77324-850 77346-514 77361-168 77384-462
5-Bromo-4-methyl-N,N-dimethylpyridine-2-carboxamide ≥95%
5-Bromo-4-methyl-N,N-dimethylpyridine-2-carboxamide

Precaución: for Research Use Only

Formula: C₉H₁₁BrN₂O
MW: 243.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28015435
CAS Number: 1809161-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR