5-Phenoxypyridin-3-ol ≥95%

Supplier: AOBChem USA

Warning

27003-500MG 27003-1G 27003-25G 27003-5G
77324-846EA 296.64 USD
77324-846 77348-690 77384-460 77372-928
5-Phenoxypyridin-3-ol ≥95%
5-Phenoxypyridin-3-ol

Precaución: for Research Use Only

Formula: C₁₁H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD20923566
CAS Number: 910649-40-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR