6-Methoxy-5-methyl-3-pyridinecarbonitrile ≥95%

Supplier: AOBChem USA

Warning

26974-500MG 26974-1G 26974-5G 26974-25G
77324-844EA 243.08 USD
77324-844 77348-688 77361-164 77384-458
6-Methoxy-5-methyl-3-pyridinecarbonitrile ≥95%
6-Methoxy-5-methyl-3-pyridinecarbonitrile

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₂O
MW: 176.215 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12024298
CAS Number: 1011476-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR