5-Chloro-6-isopropoxynaphthalene-2-boronic acid ≥97%

Supplier: AOBChem USA

Warning

26945-500MG 26945-1G 26945-5G
77339-398EA 586.07 USD
77339-398 77348-686 77372-926
5-Chloro-6-isopropoxynaphthalene-2-boronic acid ≥97%
5-Chloro-6-isopropoxynaphthalene-2-boronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₄BClO₃
Storage Temperature: Ambient
MDL Number: MFCD28142586
CAS Number: 2121511-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR