3-(Trifluoromethyl)biphenyl-4-ylboronic acid ≥95%

Supplier: AOBChem USA

Warning

26941-500MG 26941-1G 26941-5G
77324-840EA 317.24 USD
77324-840 77348-684 77372-924
3-(Trifluoromethyl)biphenyl-4-ylboronic acid ≥95%
3-(Trifluoromethyl)biphenyl-4-ylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₀BF₃O₂
MW: 266.024 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27756692
CAS Number: 2121512-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR