2-Fluoro-4-(methylthio)-3-pyridinemethanol ≥95%

Supplier: AOBChem USA

Warning

26940-500MG 26940-1G 26940-5G
77337-700EA 437.75 USD
77337-700 77348-682 77372-922
2-Fluoro-4-(methylthio)-3-pyridinemethanol ≥95%
2-Fluoro-4-(methylthio)-3-pyridinemethanol

Precaución: for Research Use Only

Formula: C₇H₈FNOS
MW: 173.208 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27756691
CAS Number: 1809161-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR