Ethyl 4-bromo-1-benzothiophene-2-carboxylate ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 4-bromobenzo[b]thiophene-2-carboxylate

269394-1G 269394-5G 269394-25G
77340-924EA 62.83 USD
77340-924 77361-160 77372-918
Ethyl 4-bromo-1-benzothiophene-2-carboxylate ≥95%
Ethyl 4-bromo-1-benzothiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₉BrO₂S
Storage Temperature: Ambient
MDL Number: MFCD23699513
CAS Number: 93103-82-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR