2-Fluoro-4-(methylsulfanyl)pyridine-3-carboxylic acid ≥95%

Supplier: AOBChem USA

Warning

26939-500MG 26939-1G 26939-5G
77335-636EA 361.53 USD
77335-636 77348-680 77372-920
2-Fluoro-4-(methylsulfanyl)pyridine-3-carboxylic acid ≥95%
2-Fluoro-4-(methylsulfanyl)pyridine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₆FNO₂S
MW: 187.191 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27756690
CAS Number: 1809161-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR