3,5-Difluoro-4-iodobenzenemethanol ≥97%

Supplier: AOBChem USA

Warning

26906-500MG 26906-1G 26906-5G 26906-25G
77324-834EA 208.06 USD
77324-834 77348-674 77361-154 77384-450
3,5-Difluoro-4-iodobenzenemethanol ≥97%
3,5-Difluoro-4-iodobenzenemethanol

Precaución: for Research Use Only

Formula: C₇H₅F₂IO
MW: 270.015 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28977238
CAS Number: 1487337-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR