3-Nitrobiphenyl-4-amine ≥97%

Supplier: AOBChem USA

Warning

26894-500MG 26894-1G 26894-5G 26894-25G
77324-832EA 122.57 USD
77324-832 77343-976 77361-152 77372-916
3-Nitrobiphenyl-4-amine ≥97%
3-Nitrobiphenyl-4-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₀N₂O₂
MW: 214.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00054606
CAS Number: 4085-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR