4-Bromo-5-methyl-1-phenyl-1H-indazole ≥95%

Supplier: AOBChem USA

Warning

26882-500MG 26882-1G 26882-5G 26882-25G
77324-826EA 154.5 USD
77324-826 77345-544 77361-148 77384-446
4-Bromo-5-methyl-1-phenyl-1H-indazole ≥95%
4-Bromo-5-methyl-1-phenyl-1H-indazole

Precaución: for Research Use Only

Formula: C₁₄H₁₁BrN₂
MW: 287.155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27756682
CAS Number: 1809168-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR