(5-Methyl-6-methylsulfanyl-pyridin-3-yl)-methanol ≥97%

Supplier: AOBChem USA

Warning

26877-500MG 26877-1G 26877-5G 26877-25G
77324-824EA 245.14 USD
77324-824 77348-668 77361-146 77384-444
(5-Methyl-6-methylsulfanyl-pyridin-3-yl)-methanol ≥97%
(5-Methyl-6-methylsulfanyl-pyridin-3-yl)-methanol

Precaución: for Research Use Only

Formula: C₈H₁₁NOS
MW: 169.244 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21087238
CAS Number: 1355179-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR