4-(4-Bromophenyl)-1,2,3,6-tetrahydro-1-(phenylmethyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

26864-500MG 26864-1G 26864-5G 26864-25G
77324-822EA 188.49 USD
77324-822 77348-666 77361-144 77384-442
4-(4-Bromophenyl)-1,2,3,6-tetrahydro-1-(phenylmethyl)pyridine ≥97%
4-(4-Bromophenyl)-1,2,3,6-tetrahydro-1-(phenylmethyl)pyridine

Precaución: for Research Use Only

Formula: C₁₈H₁₈BrN
MW: 328.246 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08443704
CAS Number: 188863-87-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR