1-Benzyl-4-(4-bromophenyl)-4-piperidinol ≥97%

Supplier: AOBChem USA

Warning

26863-500MG 26863-1G 26863-5G 26863-25G
77324-820EA 99.91 USD
77324-820 77342-758 77361-142 77372-912
1-Benzyl-4-(4-bromophenyl)-4-piperidinol ≥97%
1-Benzyl-4-(4-bromophenyl)-4-piperidinol

Precaución: for Research Use Only

Formula: C₁₈H₂₀BrNO
MW: 346.262 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01467907
CAS Number: 16332-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR