5-Chloro-2-(2,2-dimethylpropanoylamino)benzoic acid 97

Supplier: AOBChem USA

Warning

26843-500MG 26843-1G 26843-5G 26843-25G
77324-816EA 152.44 USD
77324-816 77345-444 77361-138 77384-440
5-Chloro-2-(2,2-dimethylpropanoylamino)benzoic acid 97
5-Chloro-2-(2,2-dimethylpropanoylamino)benzoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₄ClNO₃
MW: 255.697 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02032448
CAS Number: 351896-22-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR