3-Methyl-4-phenylmethoxyphenol ≥97%

Supplier: AOBChem USA

Warning

26831-500MG 26831-1G 26831-5G 26831-25G
77324-814EA 120.51 USD
77324-814 77343-670 77361-136 77372-906
3-Methyl-4-phenylmethoxyphenol ≥97%
3-Methyl-4-phenylmethoxyphenol

Precaución: for Research Use Only

Formula: C₁₄H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD11847935
CAS Number: 100927-02-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR