(6-Chloro-5-methylpyridin-3-yl)methylamine ≥97%

Supplier: AOBChem USA

Warning

26775-500MG 26775-1G 26775-25G 26775-5G
77324-810EA 298.7 USD
77324-810 77348-660 77384-436 77372-904
(6-Chloro-5-methylpyridin-3-yl)methylamine ≥97%
(6-Chloro-5-methylpyridin-3-yl)methylamine

Precaución: for Research Use Only

Formula: C₇H₉ClN₂
MW: 156.613 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18254322
CAS Number: 1256824-90-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR