1-Bromo-4-(tert-pentyl)benzene ≥97%

Supplier: AOBChem USA

Warning

26724-500MG 26724-1G 26724-25G 26724-5G
77324-808EA 300.76 USD
77324-808 77348-656 77384-434 77372-900
1-Bromo-4-(tert-pentyl)benzene ≥97%
1-Bromo-4-(tert-pentyl)benzene

Precaución: for Research Use Only

Formula: C₁₁H₁₅Br
MW: 227.141 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00028024
CAS Number: 57263-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR