tert-Butyl benzyl(5-bromo-2-methylphenyl)carbamate ≥97%

Supplier: AOBChem USA

Warning

26601-1G 26601-5G 26601-25G
77341-076EA 70.04 USD
77341-076 77361-132 77372-898
tert-Butyl benzyl(5-bromo-2-methylphenyl)carbamate ≥97%
tert-Butyl benzyl(5-bromo-2-methylphenyl)carbamate

Precaución: for Research Use Only

Formula: C₁₉H₂₂BrNO₂
MW: 376.287 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631795
CAS Number: 2364584-58-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR