2,6-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-benzenemethanol ≥97%
Supplier: AOBChem USA
|
Warning
|
26596-500MG
26596-1G
26596-5G
26596-25G
77324-804EA
223.51
USD
77324-804
77348-654
77361-128
77384-430
2,6-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-benzenemethanol ≥97%
2,6-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-benzenemethanol
Precaución: for Research Use Only
Formula:
C₁₃H₁₇BF₂O₃ MW: 270.08 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD24039809 CAS Number: 1417736-43-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...