3-Chloro-2-fluoro-5-phenylpyridine ≥97%

Supplier: AOBChem USA

Warning

26474-500MG 26474-1G 26474-5G
77335-634EA 361.53 USD
77335-634 77348-652 77372-892
3-Chloro-2-fluoro-5-phenylpyridine ≥97%
3-Chloro-2-fluoro-5-phenylpyridine

Precaución: for Research Use Only

Formula: C₁₁H₇ClFN
Storage Temperature: Ambient
MDL Number: MFCD14701632
CAS Number: 1214341-47-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR