2-Bromo-5-methylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

26436-500MG 26436-1G 26436-5G 26436-25G
77324-798EA 193.64 USD
77324-798 77348-646 77361-122 77384-428
2-Bromo-5-methylphenylboronic acid pinacol ester ≥97%
2-Bromo-5-methylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₈BBrO₂
MW: 296.996 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18383436
CAS Number: 1256781-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR