Ethyl 2-hydroxy-4-methyl-6-(2-thienyl)-1,6-dihydro-5-pyrimidinecarboxylate ≥95%

Supplier: AOBChem USA

Warning

26328-500MG 26328-1G 26328-5G 26328-25G
77324-792EA 232.78 USD
77324-792 77348-640 77361-118 77384-424
Ethyl 2-hydroxy-4-methyl-6-(2-thienyl)-1,6-dihydro-5-pyrimidinecarboxylate ≥95%
Ethyl 2-hydroxy-4-methyl-6-(2-thienyl)-1,6-dihydro-5-pyrimidinecarboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂O₃S
MW: 266.316 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00124780
CAS Number: 5948-72-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR