Ethyl 4-(3-chloro-4-fluorophenyl)-6-methyl-2-oxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate ≥95%
Supplier: AOBChem USA
26326-500MG
26326-1G
26326-5G
77336-182EA
380.07
USD
77336-182
77348-638
77372-882
Ethyl 4-(3-chloro-4-fluorophenyl)-6-methyl-2-oxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate ≥95%
Ethyl 4-(3-chloro-4-fluorophenyl)-6-methyl-2-oxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate
Precaución: for Research Use Only
Formula:
C₁₄H₁₄ClFN₂O₃ MW: 312.724 g/mol |
MDL Number:
MFCD26045273 CAS Number: 1449008-02-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...