3-Ethoxycarbonyl-4-(trifluoromethoxy)phenylboronic acid pinacol ester ≥95%

Supplier: AOBChem USA

Warning

26320-500MG 26320-1G 26320-5G
77324-790EA 341.96 USD
77324-790 77348-634 77372-878
3-Ethoxycarbonyl-4-(trifluoromethoxy)phenylboronic acid pinacol ester ≥95%
3-Ethoxycarbonyl-4-(trifluoromethoxy)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₆H₂₀BF₃O₅
Storage Temperature: Ambient
MDL Number: MFCD24040005
CAS Number: 2121515-17-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR