3-Chloro-5-cyclopropylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

26286-500MG 26286-1G 26286-5G
77339-776EA 637.57 USD
77339-776 77348-632 77372-876
3-Chloro-5-cyclopropylbenzoic acid ≥97%
3-Chloro-5-cyclopropylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₉ClO₂
Storage Temperature: Ambient
MDL Number: MFCD19439935
CAS Number: 945717-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR