1-Chloro-2-ethyl-4-(methylsulfanyl)benzene ≥97%

Supplier: AOBChem USA

Warning

26283-500MG 26283-1G 26283-5G 26283-25G
77324-788EA 177.16 USD
77324-788 77346-584 77361-116 77384-422
1-Chloro-2-ethyl-4-(methylsulfanyl)benzene ≥97%
1-Chloro-2-ethyl-4-(methylsulfanyl)benzene

Precaución: for Research Use Only

Formula: C₉H₁₁ClS
MW: 186.702 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11617792
CAS Number: 1314900-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR