2-Fluoro-5-methoxypyridin-3-ol ≥97%

Supplier: AOBChem USA

Warning

26256-500MG 26256-1G 26256-5G
77324-786EA 334.75 USD
77324-786 77348-630 77372-874
2-Fluoro-5-methoxypyridin-3-ol ≥97%
2-Fluoro-5-methoxypyridin-3-ol

Precaución: for Research Use Only

Formula: C₆H₆FNO₂
MW: 143.116 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16611056
CAS Number: 1227593-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR