(3-Bromo-5-chloropyridin-4-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

26251-1G 26251-5G 26251-25G
77341-220EA 77.25 USD
77341-220 77361-112 77372-872
(3-Bromo-5-chloropyridin-4-yl)methanol ≥97%
(3-Bromo-5-chloropyridin-4-yl)methanol

Precaución: for Research Use Only

Formula: C₆H₅BrClNO
Storage Temperature: Ambient
MDL Number: MFCD16606989
CAS Number: 1064677-18-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR