(2,5-difluoro-4-methoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

26242-1G 26242-5G 26242-25G
77341-592EA 94.76 USD
77341-592 77361-110 77372-870
(2,5-difluoro-4-methoxyphenyl)methanol ≥97%
(2,5-difluoro-4-methoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₈F₂O₂
MW: 174.145 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06246888
CAS Number: 922719-76-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR