4-Chloro-3,5-difluorobenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

26240-1G 26240-5G 26240-25G
77340-616EA 60.77 USD
77340-616 77361-108 77372-868
4-Chloro-3,5-difluorobenzyl alcohol ≥97%
4-Chloro-3,5-difluorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₅ClF₂O
MW: 178.564 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23795324
CAS Number: 1431329-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR