3-Bromo-4-(1-piperidinylmethyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

26196-500MG 26196-1G 26196-5G 26196-25G
77324-770EA 222.48 USD
77324-770 77348-618 77361-098 77384-412
3-Bromo-4-(1-piperidinylmethyl)pyridine ≥95%
3-Bromo-4-(1-piperidinylmethyl)pyridine

Precaución: for Research Use Only

Formula: C₁₁H₁₅BrN₂
MW: 255.154 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26045265
CAS Number: 1449008-24-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR