2-Amino-3-chloro-5-fluorobiphenyl ≥97%

Supplier: AOBChem USA

Warning

26187-500MG 26187-1G 26187-5G 26187-25G
77324-768EA 249.26 USD
77324-768 77348-616 77361-096 77384-410
2-Amino-3-chloro-5-fluorobiphenyl ≥97%
2-Amino-3-chloro-5-fluorobiphenyl

Precaución: for Research Use Only

Formula: C₁₂H₉ClFN
MW: 221.658 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26045263
CAS Number: 1449008-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR