4-Bromo-2,5-difluoro-N,N-dimethylbenzamide ≥95%

Supplier: AOBChem USA

Warning

26186-500MG 26186-1G 26186-5G 26186-25G
77324-766EA 222.48 USD
77324-766 77348-614 77361-094 77384-408
4-Bromo-2,5-difluoro-N,N-dimethylbenzamide ≥95%
4-Bromo-2,5-difluoro-N,N-dimethylbenzamide

Precaución: for Research Use Only

Formula: C₉H₈BrF₂NO
MW: 264.067 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26045262
CAS Number: 1449008-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR