4-(6-Fluoro-3-pyridinyl)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

26173-500MG 26173-1G 26173-5G 26173-25G
77324-764EA 218.36 USD
77324-764 77348-610 77361-092 77384-406
4-(6-Fluoro-3-pyridinyl)benzonitrile ≥97%
4-(6-Fluoro-3-pyridinyl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₂H₇FN₂
Storage Temperature: Ambient
MDL Number: MFCD32215261
CAS Number: 832735-44-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR