2-Fluoro-5-phenylpyridine-3-boronic acid ≥97%

Supplier: AOBChem USA

Warning

26172-500MG 26172-1G 26172-5G
77337-476EA 415.09 USD
77337-476 77348-608 77372-858
2-Fluoro-5-phenylpyridine-3-boronic acid ≥97%
2-Fluoro-5-phenylpyridine-3-boronic acid

Precaución: for Research Use Only

Formula: C₁₁H₉BFNO₂
Storage Temperature: Ambient
MDL Number: MFCD26045259
CAS Number: 2121514-00-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR