4-Chloro-2-fluoro-3-methoxypyridine ≥95%

Supplier: AOBChem USA

Warning

26139-500MG 26139-1G 26139-25G 26139-5G
77324-762EA 306.94 USD
77324-762 77348-604 77384-404 77372-854
4-Chloro-2-fluoro-3-methoxypyridine ≥95%
4-Chloro-2-fluoro-3-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅ClFNO
MW: 161.561 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16610378
CAS Number: 1227598-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR