4-Bromo-2-fluoro-3-methoxypyridine ≥95%

Supplier: AOBChem USA

Warning

26138-500MG 26138-1G 26138-5G
77336-860EA 396.55 USD
77336-860 77348-602 77372-852
4-Bromo-2-fluoro-3-methoxypyridine ≥95%
4-Bromo-2-fluoro-3-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅BrFNO
MW: 206.012 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16609957
CAS Number: 1227578-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR