N-(4-Bromo-3-fluorophenyl)-2,2,2-trifluoroacetamide ≥97%

Supplier: AOBChem USA

Warning

26064-500MG 26064-1G 26064-5G 26064-25G
77324-754EA 234.84 USD
77324-754 77348-598 77361-082 77384-398
N-(4-Bromo-3-fluorophenyl)-2,2,2-trifluoroacetamide ≥97%
N-(4-Bromo-3-fluorophenyl)-2,2,2-trifluoroacetamide

Precaución: for Research Use Only

Formula: C₈H₄BrF₄NO
MW: 286.021 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20807165
CAS Number: 1496430-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR