2,6-Dichloro-3-ethoxypyridine ≥97%

Supplier: AOBChem USA

Warning

26043-500MG 26043-1G 26043-5G 26043-25G
77324-750EA 199.82 USD
77324-750 77348-596 77361-074 77384-394
2,6-Dichloro-3-ethoxypyridine ≥97%
2,6-Dichloro-3-ethoxypyridine

Precaución: for Research Use Only

Formula: C₇H₇Cl₂NO
MW: 192.043 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24848697
CAS Number: 1418153-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR